Anti-SPR Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI133797EA 567 EUR
USBI133797
Anti-SPR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SPR (sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR