Anti-Nepmucin Sheep Polyclonal Antibody (Biotin)

Supplier: United States Biological
USBIN2029EA 857 EUR
USBIN2029
Anti-Nepmucin Sheep Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Nepmucin
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR