Anti-PAPPA2 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIP6000-90AEA 633 EUR
USBIP6000-90A
Anti-PAPPA2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: PAPPA2 (pappalysin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR