Anti-NR1I2 Goat Polyclonal Antibody

Supplier: United States Biological
USBIP6001-30EEA 633 EUR
USBIP6001-30E
Anti-NR1I2 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:4000; Western Blot: 1-3?g/ml; Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: NR1I2 (nuclear receptor subfamily 1, group I, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR