Anti-PRELP Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIP6201-30EA 587 EUR
USBIP6201-30
Anti-PRELP Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: PRELP (proline/arginine-rich end leucine-rich repeat protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR