Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRELP Sheep polyclonal antibody unconjugated

Supplier: US Biological
Recommended dilution: Western Blot: 1 μg/ml

Type: Primary
Antigen: PRELP (proline/arginine-rich end leucine-rich repeat protein)
Clonality: polyclonal
Conjugation: unconjugated
Host: Sheep
Isotype: IgG
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG USBIP6201-30A
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol PRELP
Antigen name proline/arginine-rich end leucine-rich repeat Protein
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Sheep
Isotype IgG
Western blot Yes
Neutralisation Yes
Amino acid number 21 to 382
Concentration ~0,1 mg/ml (after reconstitution)
Purification purified by immunoaffinity chromatography
Form lyophilized powder
Storage buffer PBS,trehalose.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty