Anti-Preproneuropeptide Y Guinea pig Polyclonal Antibody

Supplier: United States Biological
USBIP6260-03EA 381 EUR
USBIP6260-03
Anti-Preproneuropeptide Y Guinea pig Polyclonal Antibody
Antibodies
Recommended dilution: Immunochemistry: 1:500

Type: Primary
Antigen: Preproneuropeptide Y
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR