Anti-PSEN1 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIP6300-25BEA 688 EUR
USBIP6300-25B
Anti-PSEN1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:500; Immunoprecipitation: 1:100

Type: Primary
Antigen: PSEN1 (presenilin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR