Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSEN1 Rabbit polyclonal antibody Carboxyfluorescein

Supplier: US Biological
Type: Primary
Antigen: PSEN1 (presenilin 1)
Clonality: polyclonal
Conjugation: Carboxyfluorescein
Host: Rabbit
Isotype: IgG
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µl 100 µl USBIP6300-27D
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol PSEN1
Antigen name presenilin 1
Reactivity Human
Conjugation FAM (Carboxyfluorescein)
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Amino acid number 431 to 467
Purification purified
Form liquid
Storage temperature Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty