Anti-PSEN1 Rabbit polyclonal antibody

USBIP6300-27FEA 352 EUR
USBIP6300-27F
Anti-PSEN1 Rabbit polyclonal antibody
Type: Primary
Antigen: PSEN1 (presenilin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR