Anti-PSEN1 Rabbit Polyclonal Antibody (Biotin)

Supplier: United States Biological
USBIP6300-28F-BIOTEA 852 EUR
USBIP6300-28F-BIOT
Anti-PSEN1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: PSEN1 (presenilin 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR