Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSEN1 Rabbit monoclonal antibody unconjugated [clone: D39D1]

Supplier: US Biological
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: PSEN1 (presenilin 1)
Clonality: monoclonal
Clone: D39D1
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µl 100 µl USBIP6300-28L
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol PSEN1
Antigen name presenilin 1
Reactivity Monkey, Human, Mouse, Rat
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone D39D1
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoPrecipitation Yes
Format Supernatant
Form liquid
Storage buffer 10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, <0,02% sodium azide, 50% glycerol.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping