Anti-PSEN1 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIP6300-28NEA 754 EUR
USBIP6300-28N
Anti-PSEN1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: PSEN1 (presenilin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR