Anti-PSEN1 Mouse Monoclonal Antibody [clone: 6D682]

Supplier: United States Biological
USBIP6300-29FEA 983 EUR
USBIP6300-29F
Anti-PSEN1 Mouse Monoclonal Antibody [clone: 6D682]
Antibodies
Recommended dilution: ELISA: 1 ?g/ml; Immunochemistry: 6?g/ml; Western Blot: 12?g/ml

Type: Primary
Antigen: PSEN1 (presenilin 1)
Clonality: monoclonal
Clone: 6D682
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR