Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSEN2 Chicken polyclonal antibody unconjugated

Supplier: US Biological
Recommended dilution: Immunoblot/ ELISA: 1:1000 to 1:4000

Type: Primary
Antigen: PSEN2 (presenilin 2)
Clonality: polyclonal
Conjugation: unconjugated
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
500 µg 500 µG USBIP6300-41
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol PSEN2
Antigen name presenilin 2
Reactivity Mouse, Rat, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Chicken
Isotype IgY
Western blot Yes
Concentration ~1 mg/ml
Purification purified
Form liquid
Storage buffer PBS, pH 7,4, 0,02% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty