Anti-PSEN2 Mouse monoclonal antibody [clone: 6D683]

USBIP6300-47EA 656 EUR
USBIP6300-47
Anti-PSEN2 Mouse monoclonal antibody [clone: 6D683]
Type: Primary
Antigen: PSEN2 (presenilin 2)
Clonality: monoclonal
Clone: 6D683
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR