Anti-PSEN2 Rabbit polyclonal antibody

USBIP6300-47HEA 352 EUR
USBIP6300-47H
Anti-PSEN2 Rabbit polyclonal antibody
Type: Primary
Antigen: PSEN2 (presenilin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR