Anti-SRC Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS6504-45REA 755 EUR
USBIS6504-45R
Anti-SRC Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: SRC (v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR