Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NCOA3 Mouse monoclonal antibody unconjugated [clone: 8C309]

Supplier: US Biological
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: NCOA3 (nuclear receptor coactivator 3)
Clonality: Monoclonal
Clone: 8C309
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG USBIS6505-08C
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol NCOA3
Antigen name Nuclear receptor coactivator 3
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 8C309
Host Mouse
Isotype IgG1
Western blot Yes
Concentration ~0,1 mg/ml
Purification purified by Protein G chromatography
Form liquid
Storage buffer PBS, pH 7,4, 1% BSA, 0,05% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty