Anti-SRD5A1 Goat Polyclonal Antibody

Supplier: United States Biological
USBIS6505-08L5EA 551 EUR
USBIS6505-08L5
Anti-SRD5A1 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 164000; Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: SRD5A1 (steroid-5-alpha-reductase, alpha polypeptide 1 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR