Anti-SREBP-1 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS6505-16BEA 466 EUR
USBIS6505-16B
Anti-SREBP-1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: SREBP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR