Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SRF Rabbit monoclonal antibody unconjugated [clone: 11C1071]

Supplier: US Biological

Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50ChImmunoprecipitation: 1:100

Type: Primary
Antigen: SRF (serum response factor (c-fos serum response element-binding transcription factor))
Clonality: Monoclonal
Clone: 11C1071
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Pig, Rat



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µl 100 µl USBIS6560-03
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol SRF
Antigen name Serum Response Factor
Reactivity Pig, Rat, Mouse, Human, Monkey
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 11C1071
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoPrecipitation Yes
Purification purified
Form liquid
Storage Buffer 10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, less than 0,02% sodium azide, 50% glycerol.
Storage temperature Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty