Anti-SRm160 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS6567-50EA 250 EUR
USBIS6567-50
Anti-SRm160 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:162500

Type: Primary
Antigen: SRm160
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rodent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR