Anti-SRMS Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS6567-60AEA 587 EUR
USBIS6567-60A
Anti-SRMS Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: SRMS (src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR