|
|
ORDER
SPECIFICATIONS
Antigen symbol | SRPK1 |
Antigen name | SRSF protein kinase 1 |
Reactivity | Human |
Conjugation | AP (Alkaline Phosphatase) |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Epitope | N-terminal |
Purification | purified by Protein G chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,2, contains ~0,09% sodium azide |
Storage temperature | Cool |
Shipping temperature | Ice |