Anti-SRPX2 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS6570-82AEA 633 EUR
USBIS6570-82A
Anti-SRPX2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: SRPX2 (sushi-repeat containing protein, X-linked 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR