Anti-SRXN1 Goat Polyclonal Antibody

Supplier: United States Biological
USBIS6580-50EA 551 EUR
USBIS6580-50
Anti-SRXN1 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:32000; Western Blot: 0.5-1. 5 ?g/ml

Type: Primary
Antigen: SRXN1 (sulfiredoxin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR