Anti-TRIM21 Sheep Polyclonal Antibody

Supplier: United States Biological
USBIS6600-66BEA 757 EUR
USBIS6600-66B
Anti-TRIM21 Sheep Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: TRIM21 (tripartite motif containing 21)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR