Anti-ST2 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS6606-50BEA 548 EUR
USBIS6606-50B
Anti-ST2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: ST2 (suppression of tumorigenicity 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR