Anti-ST7 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS6606-57AEA 633 EUR
USBIS6606-57A
Anti-ST7 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: ST7 (suppression of tumorigenicity 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR