Anti-S Tag Mouse Monoclonal Antibody [clone: 10F401]

Supplier: United States Biological
USBIS6609-95JEA 843 EUR
USBIS6609-95J
Anti-S Tag Mouse Monoclonal Antibody [clone: 10F401]
Antibodies
Recommended dilution: Flow Cytometry:1:5;

Type: Tag
Antigen: S Tag (S tag)
Clonality: Monoclonal
Clone: 10F401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR