Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S Tag Mouse monoclonal antibody FITC (Fluorescein) [clone: 5K160]

Supplier: US Biological
Type: Tag
Antigen: S Tag (S tag)
Clonality: Monoclonal
Clone: 5K160
Conjugation: FITC (Fluorescein)
Host: Mouse
Isotype: IgG2b Kappa

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
500 µg 500 µG USBIS6609-95L
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol S
Antigen name S tag
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Monoclonal
Antibody type Tag
Clone 5K160
Host Mouse
Isotype IgG2b kappa
Flow cytometry Yes
Concentration ~0,5 mg/ml
Purification purified
Form liquid
Storage buffer PBS, 0,1% sodium azide
Storage temperature Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty