Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S Tag Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 5K160]

Supplier: US Biological
Type: Tag
Antigen: S Tag (S tag)
Clonality: Monoclonal
Clone: 5K160
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
1 ml 1 ml USBIS6609-95M
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol S
Antigen name S tag
Conjugation HRP (Horseradish Peroxidase)
Clonality Monoclonal
Antibody type Tag
Clone 5K160
Host Mouse
Isotype IgG2b kappa
ELISA Yes
Purification purified
Form liquid
Storage buffer 50% glycerol/ 50% PBS, pH 7,4
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping