Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S Tag Mouse monoclonal antibody unconjugated [clone: 10F401]

Supplier: US Biological
Recommended dilution: Flow Cytometry: 1:5-1:10;

Type: Tag
Antigen: S Tag (S tag)
Clonality: Monoclonal
Clone: 10F401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG USBIS6609-95R
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol S
Antigen name S tag
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Tag
Clone 10F401
Host Mouse
Isotype IgG2a
ELISA Yes
Flow cytometry Yes
Concentration ~0,5 mg/ml
Purification purified by immunoaffinity chromatography
Form liquid
Storage buffer borate buffered saline, pH 8.0
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty