Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S Tag Goat polyclonal antibody Agarose

Supplier: US Biological
Type: Tag
Antigen: S Tag (S tag)
Clonality: Polyclonal
Clone:
Conjugation: Agarose
Epitope:
Host: Goat
Isotype:
Reactivity:




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
1 ml 1 ml USBIS6609-96C
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol S
Antigen name S tag
Conjugation Agarose
Clonality Polyclonal
Antibody type Tag
Host Goat
ELISA Yes
Flow cytometry Yes
Purification purified by affinity chromatography
Form liquid
Storage buffer 100mM borate buffered saline, pH 8,2. No oramine- containing buffer salts .
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty