Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FUT4 Mouse monoclonal antibody unconjugated [clone: 13A269]

Supplier: US Biological
Type: Primary
Antigen: FUT4 (fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific))
Clonality: Monoclonal
Clone: 13A269
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
25 µg 25 µG USBIS6609-99
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol FUT4
Antigen name Fucosyltransferase 4
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 13A269
Host Mouse
Isotype IgM
Western blot Yes
ImmunoPrecipitation Yes
Flow cytometry Yes
Concentration ~0,5 mg/ml
Purification affinity-purified
Form liquid
Storage buffer PBS, 0,05% BSA, 0,05% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty