Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Stage Specific Embryonic Antigen 4 Mouse monoclonal antibody unconjugated [clone: 5C1]

Supplier: US Biological
Recommended dilution: Flow Cytometry: 10-20 μg/ml

Type: Primary
Antigen: Stage Specific Embryonic Antigen 4
Clonality: Monoclonal
Clone: 5C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG USBIS6610-10
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol SSEA-4
Antigen name Stage Specific Embryonic Antigen 4
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 5C1
Host Mouse
Isotype IgG3
ELISA Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Concentration ~1 mg/ml
Purification purified by Protein A chromatography
Form liquid
Storage buffer 0,02M PBS, 0,25M sodium chloride, pH 7.6, 0,1% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty