Anti-Stage Specific Embryonic Antigen 4 Mouse Monoclonal Antibody NL®557 [clone: 9L854]

Supplier: United States Biological
USBIS6610-10AEA 681 EUR
USBIS6610-10A
Anti-Stage Specific Embryonic Antigen 4 Mouse Monoclonal Antibody NL®557 [clone: 9L854]
Antibodies
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Stage Specific Embryonic Antigen 4
Clonality: Monoclonal
Clone: 9L854
Conjugation: NL®557
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR