Anti-Stage Specific Embryonic Antigen 4 Mouse monoclonal antibody FITC (Fluorescein) [clone: 11E346]

USBIS6610-10GEA 253 EUR
USBIS6610-10G
Anti-Stage Specific Embryonic Antigen 4 Mouse monoclonal antibody FITC (Fluorescein) [clone: 11E346]
Type: Primary
Antigen: Stage Specific Embryonic Antigen 4
Clonality: Monoclonal
Clone: 11E346
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Chicken, Dog, Human, Monkey, Mouse, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR