Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Stage Specific Embryonic Antigen 4 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 11E346]

Supplier: US Biological
Do not freeze!

Type: Primary
Antigen: Stage Specific Embryonic Antigen 4
Clonality: Monoclonal
Clone: 11E346
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Chicken, Dog, Human, Monkey, Mouse, Rabbit




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
25 Tests 25 Tests USBIS6610-10H
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol SSEA-4
Antigen name Stage Specific Embryonic Antigen 4
Reactivity Mouse, Chicken, Rabbit, Dog, Monkey, Human
Conjugation PE (Phycoerythrin)
Clonality Monoclonal
Antibody type Primary
Clone 11E346
Host Mouse
Isotype IgG3 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Purification purified by Protein G chromatography
Form liquid
Storage buffer PBS, 0,05% BSA, 0,05% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty