Anti-SRT Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS7049-85EA 343 EUR
USBIS7049-85
Anti-SRT Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.1-1.0 ?g/ml

Type: Tag
Antigen: SRT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR