Anti-STEAP1 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS7500-04CEA 633 EUR
USBIS7500-04C
Anti-STEAP1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: STEAP1 (six transmembrane epithelial antigen of the prostate 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR