Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STEAP1 Mouse monoclonal antibody unconjugated [clone: J2D2]

Supplier: US Biological
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 10 μg/ml

Type: Primary
Antigen: STEAP1 (six transmembrane epithelial antigen of the prostate 1)
Clonality: Monoclonal
Clone: J2D2
Conjugation: Unconjugated
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG USBIS7500-04D
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol STEAP1
Antigen name Six transmembrane epithelial antigen of the prostate 1
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone J2D2
Host Mouse
Isotype IgG2b kappa
Western blot Yes
Amino acid number 1 to 70
Purification purified
Form liquid
Storage buffer PBS, pH 7,4, 0,1% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty