Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Six-transmembrane epithelial antigen of the prostate-1 Goat polyclonal antibody unconjugated

Supplier: US Biological
Recommended dilution: ELISA: 0.5-1.0 μg/ml; Western Blot: 1.0 μg/ml

Type: Primary
Antigen: Six-transmembrane epithelial antigen of the prostate-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG USBIS7500-05
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen name Six-transmembrane epithelial antigen of the prostate-1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
Amino acid number 1 to 72
Concentration ~0,2 mg/ml
Purification purified by immunoaffinity chromatography
Form lyophilized powder
Storage buffer 0,2um sterile-filtered solution, PBS, 5% trehalose.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty