Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.

Anti-STEAP2 Rabbit polyclonal antibody unconjugated

Supplier: US Biological
Recommended dilution: Immunochemistry: 10 μg/ml

Type: Primary
Antigen: STEAP2 (STEAP family member 2, metalloreductase)
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG USBIS7500-15A
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol STEAP2
Antigen name STEAP family member 2, metalloreductase
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Amino acid number 455 to 467
Concentration ~1 mg/ml
Purification purified by immunoaffinity chromatography
Form liquid
Storage buffer PBS, 0,02% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty