Anti-STEAP3 NT Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS7500-18EA 633 EUR
USBIS7500-18
Anti-STEAP3 NT Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: STEAP3 NT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR