Anti-STEAP4 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIS7500-21DEA 633 EUR
USBIS7500-21D
Anti-STEAP4 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:20000; Immunofluorescence: 1:100-1:500; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: STEAP4 (STEAP family member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR