Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SCD Goat polyclonal antibody unconjugated

Supplier: US Biological
Recommended dilution: ELISA: 1:64000; Immunochemistry: 5 μg/ml

Type: Primary
Antigen: SCD (stearoyl-CoA desaturase (delta-9-desaturase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG USBIS7900-02A
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol SCD
Antigen name Stearoyl-CoA desaturase
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Goat
ELISA Yes
Epitope C-terminal
Concentration ~0,5 mg/ml
Purification purified by immunoaffinity chromatography
Sequence C-RIKRTGDGNYKSG
Form liquid
Storage buffer Tris saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty