|
![]() |
ORDER
SPECIFICATIONS
Antigen symbol | IgG + IgM |
Antigen name | Immunoglobulin G And M |
Reactivity | Mouse |
Conjugation | HRP (Horseradish Peroxidase) |
Clonality | Polyclonal |
Antibody type | Secondary |
Host | Goat |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
ImmunoFluorescence | Yes |
Gel shift | Yes |
Flow cytometry | Yes |
Concentration | 1,0 mg/ml (E 1% at 280 nm = 13,0) |
Format | Purified IgG |
Cross adsorption | No |
Immunogen | Mouse IgG + IgM |
Purification | Affinity purified using solid phaseMouse IgG + IgM |
Form | Lyophilized |
Storage buffer | 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free |
Storage temperature | 2-8 °C |
Shipping temperature | 2-8 °C |