Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Mouse IgM Polyclonal Antibody (DyLight® 800)

Supplier: ImmunoReagents
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Goat
Isotype: IgM
Reactivity: Mouse



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
1,0 mg 1 mg IMMRIR2046
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgM
Antigen name Immunoglobulin M
Reactivity Mouse
Conjugation DyLight® 800
Clonality Polyclonal
Antibody type Secondary
Host Goat
Isotype IgM
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Concentration 1,0 mg/ml (E 1% at 280 nm = 13,0)
Format Purified IgG
Cross Adsorption No
Immunogen Mouse IgM
Purification Affinity purified using solid phaseMouse IgM
Form Lyophilized
Storage Buffer 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
Storage temperature 2-8 °C
Shipping Temperature 2-8 °C
Tested Applications Fluoresence ELISA, Fluorescence-Activated Cell Sorting
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty